Triphenylacetic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

T81205-10G T81205-5G
SIALT81205-10GEA 121 EUR
SIALT81205-10G SIALT81205-5G
Triphenylacetic acid, Sigma-Aldrich®
Triphenylacetic acid
Formula: C₂₀H₁₆O₂
Boiling Pt: 437 °C (1013 hPa)
Melting Pt: 267…273 °C
Density: 1.27 g/cm³ (20 °C)
MDL Number: MFCD00004185
CAS Number: 595-91-5
EINECS: 209-873-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR