(4-Methoxyphenyl)acetic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-Methoxyphenylacetic acid
M-Clarity™ quality level = MQ100
M19201-100G M19201-5G
SIALM19201-100GEA 62.5 EUR
SIALM19201-100G SIALM19201-5G
(4-Methoxyphenyl)acetic acid, Sigma-Aldrich®
(4-Methoxyphenyl)acetic acid
Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 306
Melting Pt: 84…86 °C
Flash Pt: 193
Storage Temperature: Ambient
MDL Number: MFCD00004345
CAS Number: 104-01-8
EINECS: 203-166-4