HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid), Sigma-Aldrich®
Supplier: Merck
Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid
54457-10G-F 54457-250G-F 54457-50G-F
SIAL54457-10G-FEA 48.3 EUR
SIAL54457-10G-F SIAL54457-250G-F SIAL54457-50G-F
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid), Sigma-Aldrich®
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
Formula: C₈H₁₈N₂O₄S
Melting Pt: 210…215 °C
Density: 1.448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM