4-Bromo-1-butene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
167851-10G 167851-1G 167851-50G
SIAL167851-10GEA 103 EUR
SIAL167851-10G SIAL167851-1G SIAL167851-50G
4-Bromo-1-butene, Sigma-Aldrich®
4-Bromo-1-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 98
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 9
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1992
ADR: 3 II