trans-2-Pentenal, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Trans-2-pentenal, trans-2-Pentenal, (E)-2-Pentenal

M-Clarity™ quality level = MQ100

269255-25G 269255-5G
SIAL269255-25GEA 111 EUR
SIAL269255-25G SIAL269255-5G
trans-2-Pentenal, Sigma-Aldrich®
trans-2-Pentenal
Formula: C₅H₈O
Boiling Pt: 127 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009615
CAS Number: 1576-87-0
EINECS: 216-414-1
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR