trans-2-Pentenal, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Trans-2-pentenal, trans-2-Pentenal, (E)-2-Pentenal
M-Clarity™ quality level = MQ300
W321818-100G-K
W321818-1KG-K
W321818-25G-K
SIALW321818-100G-KEA
109
EUR
SIALW321818-100G-K
SIALW321818-1KG-K
SIALW321818-25G-K
trans-2-Pentenal, Sigma-Aldrich®
trans-2-Pentenal
Formula:
C₅H₈O Boiling Pt: 127 °C (1013 hPa) Density: 0.86 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00009615 CAS Number: 1576-87-0 EINECS: 216-414-1 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...