3-Chloropropene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Chloro-1-propene, Allyl chloride
M-Clarity™ quality level = MQ100
236306-100ML 236306-5ML
SIAL236306-100MLEA 55.9 EUR
SIAL236306-100ML SIAL236306-5ML
3-Chloropropene, Sigma-Aldrich®
3-Chloropropene
Formula: C₃H₅Cl
MW: 76.53 g/mol
Boiling Pt: 44
Melting Pt: –136 °C
Density: 0.93 g/cm³ (20 °C)
Flash Pt: –32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000984
CAS Number: 107-05-1
EINECS: 203-457-6
UN: 1100
ADR: 3 I
Merck Index: 13,00286