Trichloroethylene, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
1,1,2-Trichloroethene, Trichloroethene, 1,1,2-Trichloroethylene
M-Clarity™ quality level = MQ100
372145-100ML
372145-1L
SIAL372145-100MLEA
50.5
EUR
SIAL372145-100ML
SIAL372145-1L
Trichloroethylene, Sigma-Aldrich®
Trichloroethylene
Formula:
ClCH=CCl₂ Boiling Pt: 86.7 °C (1013 hPa) Melting Pt: –73 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000838 CAS Number: 79-01-6 EINECS: 201-167-4 UN: 1710 ADR: 6.1,III Merck Index: 12,09769 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...