Oleic acid tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
(9Z)-9-Octadecenoic acid
A16663.AU
A16663.0J
A16663.AUEA
50.7
EUR
A16663.AU
A16663.0J
Oleic acid tech. 90%, Technical Grade
Oleic acid
Purity: 90%
CAS No.: 112-80-1
Molecular Formula: C18H34O2
Formula Weight: 282.47
Storage Temperature: 2°C to 8°C
Physical Form: Clear liquid
Density: 0.887
Refractive Index: 1.4610
Sensitivity: Air Sensitive
Appearance: Colourless
Melting Point: 13-14°
Boiling Point: 286°/100mm
Beilstein Reference: 1726542
Merck Reference: 14,6828
MDL No.: MFCD00064242
Formula:
C₁₈H₃₄O₂ Boiling Pt: 286 °C (100 mmHg) Melting Pt: 13…14 °C Density: 0.887 Flash Pt: 180 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064242 CAS Number: 112-80-1 EINECS: 204-007-1 Merck Index: 14,6828 |