Maleic anhydride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98+%
CAS No.: 108-31-6
Molecular Formula: C4H2O3
Formula Weight: 98.06
Flash Point: 103°(217°F)
Melting Point: 52-56°
Boiling Point: 202°
Beilstein Reference: 106909
Merck Reference: 14,5704
UN No.: 2215
MDL No.: MFCD00005518
Formula:
C₄H₂O₃ Boiling Pt: 202 °C Melting Pt: 52…56 °C Density: 1.48 g/cm³ (20 °C) Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005518 CAS Number: 108-31-6 EINECS: 203-571-6 UN: 2215 ADR: 8,III Merck Index: 14,5704 |