Maleic anhydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2,5-Furanedione
M-Clarity™ quality level = MQ200
M188-1KG-A M188-25G-A M188-25KG-A M188-5KG-A
SIALM188-1KG-AEA 83.2 EUR
SIALM188-1KG-A SIALM188-25G-A SIALM188-25KG-A SIALM188-5KG-A
Maleic anhydride, Sigma-Aldrich®
Maleic anhydride
Formula: C₄H₂O₃
MW: 98.06 g/mol
Boiling Pt: 200
Melting Pt: 51…53 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 103
Storage Temperature: Ambient
MDL Number: MFCD00005518
CAS Number: 108-31-6
EINECS: 203-571-6
UN: 2215
ADR: 8 III
Merck Index: 13,05727
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e48fcfd091622f49001009b383d7d62edf3521f5&vid=ca4d52f333124f172be23afe0d6384d67fc57a66&prid=web&clid=SAPDAM