Citraconic anhydride ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 616-02-4
Molecular Formula: C5H4O3
Formula Weight: 112.08
Flash Point: 101°(213°F)
Refractive Index: 1.4710
Melting Point: 7-8°
Boiling Point: 213-214°
Beilstein Reference: 1835
UN No.: 2810
MDL No.: MFCD00005522
Formula:
C₅H₄O₃ Boiling Pt: 213…214 °C Melting Pt: 7…8 °C Density: 1.243 Flash Pt: 101 °C (213 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005522 CAS Number: 616-02-4 EINECS: 210-459-0 UN: 2810 ADR: 6.1,III |