5-Hexen-1-ol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Hexen-6-ol, 5-Hexen-1-ol
M-Clarity™ quality level = MQ100
230324-25G 230324-5G
SIAL230324-25GEA 200 EUR
SIAL230324-25G SIAL230324-5G
5-Hexen-1-ol, Sigma-Aldrich®
5-Hexen-1-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 152…155 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.84 g/cm³ (20 °C)
Flash Pt: 47
Storage Temperature: Ambient
MDL Number: MFCD00002981
CAS Number: 821-41-0
EINECS: 212-477-4
UN: 1987
ADR: 3 III