trans-2-Hexen-1-ol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W256218-1KG-K W256218-250G-K W256218-8KG-K
SIALW256218-1KG-KEA 217 EUR
SIALW256218-1KG-K SIALW256218-250G-K SIALW256218-8KG-K
trans-2-Hexen-1-ol, Sigma-Aldrich®
trans-2-Hexen-1-ol
Formula: C₆H₁₂O
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: 65…67 °C
Density: 0.8459 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002927
CAS Number: 928-95-0
EINECS: 213-191-2
UN: 1987
ADR: 3,III