3-Chloro-2-methylpropene, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

108030-100ML 108030-1L
SIAL108030-100MLEA 44.4 EUR
SIAL108030-100ML SIAL108030-1L
3-Chloro-2-methylpropene, Sigma-Aldrich®
3-Chloro-2-methylpropene
Formula: C₄H₇Cl
Boiling Pt: 72 °C (1013 hPa)
Melting Pt: –80 °C
Density: 0.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000953
CAS Number: 563-47-3
EINECS: 209-251-2
UN: 2554
ADR: 3,II
Merck Index: 13,02167

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR