Cinnamyl alcohol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cinnamic alcohol, 3-Phenylprop-2-en-1-ol
M-Clarity™ quality level = MQ300
W229407-10KG-K W229407-1KG-K W229407-25KG-K W229407-5KG-K
SIALW229407-10KG-KEA 397 EUR
SIALW229407-10KG-K SIALW229407-1KG-K SIALW229407-25KG-K SIALW229407-5KG-K
Cinnamyl alcohol, Sigma-Aldrich®
Cinnamyl alcohol
Formula: C₉H₁₀O
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 31…34 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002921
CAS Number: 104-54-1
EINECS: 203-212-3
Merck Index: 13,02325