9-Bromoanthracene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

B56609-25G B56609-5G
SIALB56609-25GEA 265 EUR
SIALB56609-25G SIALB56609-5G
9-Bromoanthracene, Sigma-Aldrich®
9-Bromoanthracene
Formula: C₁₄H₉Br
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR