9-Nitroanthracene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

N10209-1G N10209-5G
SIALN10209-1GEA 49.1 EUR
SIALN10209-1G SIALN10209-5G
9-Nitroanthracene, Sigma-Aldrich®
9-Nitroanthracene
Formula: C₁₄H₉NO₂
Boiling Pt: 403 °C (1013 hPa)
Melting Pt: 137…139 °C
Density: 1.591 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001248
CAS Number: 602-60-8
EINECS: 210-021-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR