9-Methylanthracene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
M29606-10G M29606-1G
SIALM29606-10GEA 259 EUR
SIALM29606-10G SIALM29606-1G
9-Methylanthracene, Sigma-Aldrich®
9-Methylanthracene
Formula: C₁₅H₁₂
MW: 192.26 g/mol
Boiling Pt: 347
Melting Pt: 73…76 °C
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001261
CAS Number: 779-02-2
EINECS: 212-299-7
UN: 3077
ADR: 9 III