2-Piperidone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: δ-Valerolactame, delta-Valerolactam, Piperidin-2-one
M-Clarity™ quality level = MQ200
V209-100G V209-25G
SIALV209-100GEA 200 EUR
SIALV209-100G SIALV209-25G
2-Piperidone, Sigma-Aldrich®
2-Piperidone
Formula: C₅H₉NO
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 35…38 °C
Density: 1.073 g/cm³ (40 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006037
CAS Number: 675-20-7
EINECS: 211-622-9