N-Iodosuccinimide 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-Iodopyrrolidine-2,5-dione
299570100 299570500 299571000 299575000
ACRO299570100EA 116 EUR
ACRO299570100 ACRO299570500 ACRO299571000 ACRO299575000
N-Iodosuccinimide 98%
N-Iodosuccinimide
Formula: C₄H₄INO₂
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 197…204 °C
Density: 2,245 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005512
CAS Number: 516-12-1
EINECS: 208-221-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,05065
Order Now
Specification Test Results
Appearance White to yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 204 °C
Titration Iodometric ≥97.5 %