N-Iodosuccinimide
Supplier: Molekula
|
Warning
|
23668371-25G
23668371-100G
MOLE23668371-25GEA
88.5
EUR
MOLE23668371-25G
MOLE23668371-100G
N-Iodosuccinimide
N-Iodosuccinimide
Formula:
C₄H₄INO₂ Boiling Pt: 356 °C (1013 hPa) Melting Pt: 200…202 °C Density: 2.245 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005512 CAS Number: 516-12-1 EINECS: 208-221-6 Merck Index: 13,05065 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...