N-Iodosuccinimide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 1-Iodopyrrolidine-2,5-dione

M-Clarity™ quality level = MQ200

220051-100G 220051-25G 220051-5G
SIAL220051-100GEA 369 EUR
SIAL220051-100G SIAL220051-25G SIAL220051-5G
N-Iodosuccinimide, Sigma-Aldrich®
N-Iodosuccinimide
Formula: C₄H₄INO₂
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 200…202 °C
Density: 2.245 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005512
CAS Number: 516-12-1
EINECS: 208-221-6
Merck Index: 13,05065

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR