Cycloheximide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
M-Clarity™ quality level = MQ200
01810-1G 01810-5G
SIAL01810-1GEA 105 EUR
SIAL01810-1G SIAL01810-5G
Cycloheximide, Sigma-Aldrich®
Cycloheximide
Formula: C₁₅H₂₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00082346
CAS Number: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM