4-Methylmorpholine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: NMM, N-Methylmorpholine
67869-100ML 67869-25ML 67869-500ML
SIAL67869-100MLEA 153 EUR
SIAL67869-100ML SIAL67869-25ML SIAL67869-500ML
4-Methylmorpholine, Sigma-Aldrich®
4-Methylmorpholine
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 115
Melting Pt: –66 °C
Density: 0.922 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00006175
CAS Number: 109-02-4
EINECS: 203-640-0
UN: 2535
ADR: 3 II