4-Methylmorpholine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: NMM, N-Methylmorpholine

M-Clarity™ quality level = MQ200

M56557-100ML M56557-500ML
SIALM56557-100MLEA 30.9 EUR
SIALM56557-100ML SIALM56557-500ML
4-Methylmorpholine, Sigma-Aldrich®
4-Methylmorpholine
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 115
Melting Pt: –66 °C
Density: 0.922 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00006175
CAS Number: 109-02-4
EINECS: 203-640-0
UN: 2535
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR