Azulene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
A97203-100MG
A97203-1G
SIALA97203-100MGEA
71.5
EUR
SIALA97203-100MG
SIALA97203-1G
Azulene, Sigma-Aldrich®
Azulene
Formula:
C₁₀H₈ Boiling Pt: 242 °C (1013 hPa) Melting Pt: 96…97 °C |
MDL Number:
MFCD00003810 CAS Number: 275-51-4 EINECS: 205-993-6 UN: 3077 ADR: 9,III Merck Index: 13,00928 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...