2-Indolinone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Oxindole, Oxindole, Indolin-2-one, 2,3-Dihydro-2-indolone

M-Clarity™ quality level = MQ100

O9808-25G O9808-5G
SIALO9808-25GEA 65.2 EUR
SIALO9808-25G SIALO9808-5G
2-Indolinone, Sigma-Aldrich®
2-Indolinone
Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 313
Melting Pt: 122…125 °C
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00005711
CAS Number: 59-48-3
EINECS: 200-429-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR