1,2-Dihydronaphthalene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
D105937-10G
D105937-1G
D105937-25G
SIALD105937-10GEA
365
EUR
SIALD105937-10G
SIALD105937-1G
SIALD105937-25G
1,2-Dihydronaphthalene, Sigma-Aldrich®
1,2-Dihydronaphthalene
Formula:
C₁₀H₁₀ Boiling Pt: 204 °C (1013 hPa) Melting Pt: –8 °C Density: 1.0031 g/cm³ (10.5 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001672 CAS Number: 447-53-0 EINECS: 207-183-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...