4-Ethoxyphenylboronic acid ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B23683.03
B23683.06
B23683.14
B23683.03EA
47.4
EUR
B23683.03
B23683.06
B23683.14
4-Ethoxyphenylboronic acid ≥98%
4-Ethoxyphenylboronic acid
Purity: 98%
CAS No.: 22237-13-4
Molecular Formula: C8H11BO3
Formula Weight: 165.99
Melting Point: 165-168°
Beilstein Reference: 3530408
Formula:
C₈H₁₁BO₃ Melting Pt: 165…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00674028 CAS Number: 22237-13-4 |