1,2-Dibromobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
D39002-100G D39002-25G D39002-5G
SIALD39002-100GEA 109 EUR
SIALD39002-100G SIALD39002-25G SIALD39002-5G
1,2-Dibromobenzene, Sigma-Aldrich®
1,2-Dibromobenzene
Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224
Density: 1.956 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3
UN: 3082
ADR: 9 III