Bromobenzene, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenylbromide, Phenyl bromide
M-Clarity™ quality level = MQ200
B57702-100ML B57702-2L B57702-500ML B57702-18L
SIALB57702-100MLEA 19.4 EUR
SIALB57702-100ML SIALB57702-2L SIALB57702-500ML SIALB57702-18L
Bromobenzene, Sigma-Aldrich®
Bromobenzene
Formula: C₆H₅Br
MW: 157.01 g/mol
Boiling Pt: 155
Melting Pt: –31 °C
Density: 1.495 g/cm³ (20 °C)
Flash Pt: 51
Storage Temperature: Ambient
MDL Number: MFCD00000055
CAS Number: 108-86-1
EINECS: 203-623-8
UN: 2514
ADR: 3 III
Merck Index: 13,01390