1-Bromo-2-chlorobenzene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Bromochlorobenzene
M-Clarity™ quality level = MQ200
B60401-100G B60401-10G
SIALB60401-100GEA 91 EUR
SIALB60401-100G SIALB60401-10G
1-Bromo-2-chlorobenzene, Sigma-Aldrich®
1-Bromo-2-chlorobenzene
Formula: C₆H₄BrCl
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: –12.1 °C
Density: 1.65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1