1-Bromo-4-chlorobenzene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Bromochlorobenzene
M-Clarity™ quality level = MQ200
B60428-100G B60428-1KG B60428-500G
SIALB60428-100GEA 33.4 EUR
SIALB60428-100G SIALB60428-1KG SIALB60428-500G
1-Bromo-4-chlorobenzene, Sigma-Aldrich®
1-Bromo-4-chlorobenzene
Formula: C₆H₄BrCl
Boiling Pt: 196 °C (1013 hPa)
Melting Pt: 66…68 °C
Density: 1.576 g/cm³ (71 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000600
CAS Number: 106-39-8
EINECS: 203-392-3
UN: 2811
ADR: 6.1,III