4-Bromo-2-chlorophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
167789-25G 167789-5G
SIAL167789-25GEA 137 EUR
SIAL167789-25G SIAL167789-5G
4-Bromo-2-chlorophenol, Sigma-Aldrich®
4-Bromo-2-chlorophenol
Formula: C₆H₄BrClO
MW: 207.45 g/mol
Boiling Pt: 232
Melting Pt: 50…51 °C
Density: 1.617 g/cm³ (20 °C)
MDL Number: MFCD00002166
CAS Number: 3964-56-5
EINECS: 223-572-5
UN: 3082
ADR: 9 III