4-Bromobenzonitrile, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
B58407-10G B58407-50G
SIALB58407-10GEA 50.8 EUR
SIALB58407-10G SIALB58407-50G
4-Bromobenzonitrile, Sigma-Aldrich®
4-Bromobenzonitrile
Formula: C₇H₄BrN
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: ∼90 °C
Density: 1.562 g/cm³ (20 °C)
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00001811
CAS Number: 623-00-7
EINECS: 210-764-9
UN: 3439
ADR: 6.1,III