2-Bromobenzoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Bromobenzene-1-carbonyl chloride, o-Bromobenzoyl chloride
M-Clarity™ quality level = MQ100
B59179-10G B59179-50G
SIALB59179-10GEA 58.4 EUR
SIALB59179-10G SIALB59179-50G
2-Bromobenzoyl chloride, Sigma-Aldrich®
2-Bromobenzoyl chloride
Formula: C₇H₄BrClO
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 11 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000655
CAS Number: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8,II