1-Bromo-4-fluorobenzene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
B67201-100G B67201-25G B67201-500G
SIALB67201-100GEA 54.4 EUR
SIALB67201-100G SIALB67201-25G SIALB67201-500G
1-Bromo-4-fluorobenzene, Sigma-Aldrich®
1-Bromo-4-fluorobenzene
Formula: C₆H₄BrF
Boiling Pt: 151 °C (1013 hPa)
Melting Pt: –16 °C
Density: 1.4946 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00000342
CAS Number: 460-00-4
EINECS: 207-300-2
UN: 1993
ADR: 3,III