1-Bromo-2-(trifluoromethoxy)benzene
Supplier: Thermo Scientific
TL00649.1
TL00649.10
TL00649.50
MAYBTL00649.1EA
0
EUR
MAYBTL00649.1
MAYBTL00649.10
MAYBTL00649.50
1-Bromo-2-(trifluoromethoxy)benzene
1-Bromo-2-(trifluoromethoxy)benzene
Formula:
C₇H₄BrF₃O |
MDL Number:
MFCD00040942 CAS Number: 64115-88-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...