2-Bromo-3,5-difluorophenol ≥98%
Supplier: Thermo Fisher Scientific
Warning
H26203.03 H26203.06
H26203.03EA 53.5 EUR
H26203.03 H26203.06
2-Bromo-3,5-difluorophenol ≥98%
2-Bromo-3,5-difluorophenol
Purity: 98% CAS No.: 325486-43-9 Molecular Formula: C6H3BrF2O Formula Weight: 208.99 Refractive Index: 1.529
Formula: C₆H₃BrF₂O
MW: 208.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861127
CAS Number: 325486-43-9
EINECS: None