2-Bromothiophenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
242659-10G
242659-50G
SIAL242659-10GEA
107
EUR
SIAL242659-10G
SIAL242659-50G
2-Bromothiophenol, Sigma-Aldrich®
2-Bromothiophenol
Formula:
C₆H₅BrS Boiling Pt: 128 °C (1013 hPa) Density: 1.604 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00004827 CAS Number: 6320-02-1 EINECS: 228-665-4 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...