2-Bromobenzenesulphonyl chloride

Supplier: Thermo Scientific

Synonyms: 2-Bromobenzene-1-sulphonyl chloride

TL00534.1 TL00534.10 TL00534.50
MAYBTL00534.1EA 0 EUR
MAYBTL00534.1 MAYBTL00534.10 MAYBTL00534.50
2-Bromobenzenesulphonyl chloride
2-Bromobenzenesulphonyl chloride
Formula: C₆H₄BrClO₂S
Boiling Pt: 127…128 °C (3.6 mmHg)
Melting Pt: 51 °C
MDL Number: MFCD00051971
CAS Number: 2905-25-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR