Benzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ300

W212709-25KG-K W212709-10KG-K W212709-1KG-K
SIALW212709-25KG-KEA 384 EUR
SIALW212709-25KG-K SIALW212709-10KG-K SIALW212709-1KG-K
Benzaldehyde, Sigma-Aldrich®
Benzaldehyde
Formula: C₆H₅CHO
MW: 106.12 g/mol
Boiling Pt: 179
Melting Pt: –26 °C
Density: 1.049 g/cm³ (20 °C)
Flash Pt: 64
Storage Temperature: Ambient
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9 III
Merck Index: 13,01057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98811dd8cad9340872b5c4b5bfb305860eb1f81a&vid=934c1b87225e77b213e66c334d8f27ef4d924847&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR