Vanillic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
W398802-10KG-K W398802-1KG-K
SIALW398802-10KG-KEA 3970 EUR
SIALW398802-10KG-K SIALW398802-1KG-K
Vanillic acid, Sigma-Aldrich®
Vanillic acid
Formula: C₈H₈O₄
Boiling Pt: 353 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Merck Index: 12,10068