4-(Dimethylamino)benzaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
M-Clarity™ quality level = MQ100
109762-100G
109762-500G
SIAL109762-100GEA
55.5
EUR
SIAL109762-100G
SIAL109762-500G
4-(Dimethylamino)benzaldehyde, Sigma-Aldrich®
4-(Dimethylamino)benzaldehyde
Formula:
(CH₃)₂NC₆H₄CHO Boiling Pt: 176…177 °C (1013 hPa) Melting Pt: 72 °C Density: 1.0254 g/cm³ (99.9 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003381 CAS Number: 100-10-7 EINECS: 202-819-0 Merck Index: 13,03257 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...