4-Methoxysalicylaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Hydroxy-4-methoxybenzaldehyde
M-Clarity™ quality level = MQ100
160695-25G 160695-5G
SIAL160695-25GEA 520 EUR
SIAL160695-25G SIAL160695-5G
4-Methoxysalicylaldehyde, Sigma-Aldrich®
4-Methoxysalicylaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 124 °C (12 mmHg)
Melting Pt: 42…43 °C
Density: 1.2065 g/cm³ (51 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003327
CAS Number: 673-22-3
EINECS: 211-604-0