Ethylvanillin, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W246409-10KG-K W246409-1KG-K
SIALW246409-10KG-KEA 590 EUR
SIALW246409-10KG-K SIALW246409-1KG-K
Ethylvanillin, Sigma-Aldrich®
Ethylvanillin
Formula: C₉H₁₀O₃
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 75…77 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006944
CAS Number: 121-32-4
EINECS: 204-464-7
Merck Index: 13,03890

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR