2,5-Dihydroxybenzaldehyde, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
D108200-25G
D108200-5G
SIALD108200-25GEA
688
EUR
SIALD108200-25G
SIALD108200-5G
2,5-Dihydroxybenzaldehyde, Sigma-Aldrich®
2,5-Dihydroxybenzaldehyde
Formula:
C₇H₆O₃ MW: 138.12 g/mol Boiling Pt: 60…65 °C (12 mmHg) Melting Pt: 99 Density: 0.859 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003333 CAS Number: 1194-98-5 EINECS: 214-789-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...