Chlorobenzene, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Phenylchloride, Benzene Chloride, Monochlorobenzene

M-Clarity™ quality level = MQ100

284513-100ML 284513-1L
SIAL284513-100MLEA 107 EUR
SIAL284513-100ML SIAL284513-1L
Chlorobenzene, Sigma-Aldrich®
Chlorobenzene
Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 130
Melting Pt: –45 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3 III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3de7b2cdf345bec36e545333b590a8be006958f&vid=feb76aea4ef0ddc6c13a4844dc1ea1a8751259db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR