3-Chlorophenylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
B24444.06
B24444.03
B24444.14
B24444.06EA
73
EUR
B24444.06
B24444.03
B24444.14
3-Chlorophenylboronic acid ≥97%
3-Chlorophenylboronic acid
Purity: 97%
CAS No.: 63503-60-6
Molecular Formula: C6H6BClO2
Formula Weight: 156.38
Melting Point: 180-185°
Beilstein Reference: 2936343
Formula:
C₆H₆BClO₂ Melting Pt: 180…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161354 CAS Number: 63503-60-6 |